missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Congo paper MN 260 HE pH Paper
Shop All Macherey-Nagel Products
Click to view available options
Color Indicator:
Red to Blue
Unit Size:
Pack of 5
Specifications
Specifications
| Color Indicator | Red to Blue |
| pH Range | 5 to 3 |
| Format | Strip |
| For Use With (Application) | Determination of lactic acid |
| Quantity | 200 Pack |
| Type | pH Paper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction