Learn More
Lenz Laborglasintrumente Cones
Description
- Acc. to DIN 12249 form B and in addition to standard
- Conical ground cones
- Made of DURAN™ tubing
Specifications
Specifications
| Quantity | 1 Ea. |
| Joint Size | NS 60/46 |
| Thickness (Metric) Wall | 2.5 mm |
| Diameter (Metric) Outer Tube | 54 mm |
| Material | Borosilicate Glass (3.3) |