Learn More
Brady™ Cone™ Weather-resistant Blank Cones
Description
- Made of high impact, weather-resistant materials.
- Ideal for your indoor or outdoor hazardous warning needs.
- Cones are four-sided, so your message can't be missed.
Specifications
Specifications
| Alternate Part Number | CONE-4SIDE-35H-ORANGE-BLANK |
| Displayed Text | Blank |
| Height (English) | 35 in. |
| Height (Metric) | 889 mm |
| Length (English) | 10.5 in. |
| Length (Metric) | 266.7 mm |
| Packaging Quantity | 1 Ea. |
| Color | Orange |
| Material | Plastic |
| Type | Blank Cone |