Learn More
Quickfit™ Single Cone with Long Tip
Product Code. 15587760
Change view
Click to view available options
Cone Size:
14/23
19/26
24/29
29/32
34/35
40/38
43027
45/40
55/44
7/16
Thickness (Metric) Wall:
1mm
1.5mm
1.8mm
2mm
2.3mm
2.5mm
Unit Size:
Pack of 10
Pack of 50
Product Code. Cone Size Thickness (Metric) Wall unitSize
15587760 55/44 2.5mm Pack of 50
12413581 14/23 1.5mm Pack of 10
12493581 7/16 1.5mm Pack of 10
15547760 43027 1mm Pack of 50
15567760 40/38 2mm Pack of 50
15577760 45/40 2.3mm Pack of 50
12423581 19/26 1.8mm Pack of 10
12433581 24/29 2mm Pack of 10
12443581 29/32 2mm Pack of 10
15557760 34/35 2mm Pack of 50
10 options
Product Code. 15587760
Brand: Quickfit CBB55UB
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Allows cone to be joined to glass tubing
  • Useful for construction of stirrer guides
  • Unbadged and free from ceramic markings
  • Avoids problem of badge charring at high temperatures which can give an untidy appearance
  • Long tip cones manufactured with minimum shank length of 90mm
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10/Pk.
Item Description Dia. Shank: 52mm, Bore: 40mm
Type Single Cone with Long Tip
Cone Size 55/44
Thickness (Metric) Wall 2.5mm