Learn More
Conductivity Test Solution Potassium Chloride, SI Analytics™
Shop All SI Analytics Products
Click to view available options
Quantity:
20 mL
250 mL
500 mL
Packaging:
3 x 6 Ampules
Polyethylene Bottle
Concentration:
0.01 mol/L
0.01/0.1/1 mol/L (1.41/12.9/112mS/cm)
5mS/cm
Unit Size:
250mL
Each
Description
- Mixture consisting of water and potassium chloride
- Store at room temperature
- The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
- Product does not present an explosion hazard
- Do not allow undiluted product or large quantities of it to reach ground water, water course or sewage system
- Small amounts may be diluted with plenty of water and washed away
Specifications
Specifications
| Packaging | 3 x 6 Ampules |
| Odor | Odorless |
| Density | 1g/cm3 |
| Physical Form | Fluid |
| Concentration | 0.01/0.1/1 mol/L (1.41/12.9/112mS/cm) |
| Color | Colorless |
| pH | 7 |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Solubility | Miscible with water |
| Show More |