Learn More
Conductivity Standards 500μS/cm, Mettler Toledo™
Product Code. 11977501
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11977501 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11977501 Supplier Mettler Toledo Online Supplier No. 51300170
  • ONLINE EXCLUSIVE PRICE
    1325.00 NOK
    455.21 NOK / 250mL
    Save 869.79 NOK
    65% Off
 Restricted Item
In Stock
Specifications
Description Cond standard 500 µs/cm, 250 mL
Packaging 250 mL
Conductivity 500μS/cm
Quantity 250 mL