Learn More
Conductivity Standard, 500μS/cm, NIST Certified, Chem-Lab
Product Code. 11903174
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 11903174
Brand: Chem-Lab CL03.1121.0500
Calibrates electrodes and meters for accurate electrical and thermal conductivity measurements. Conductivity Standard, 500μS/cm, NIST Certified, Chem-Lab is a potassium chloride solution for laboratory use and LF measurements. 452μS at 20°C, 500μS at 25°C (±1%).
15°C—402.9μS/cm
16°C—412.6μS/cm
17°C—422.4μS/cm
18°C—432.1μS/cm
19°C—441.8μS/cm
20°C—451.5μS/cm
21°C—461.2μS/cm
22°C—470.9μS/cm
23°C—480.6μS/cm
24°C—490.3μS/cm
25°C—500.0μS/cm
30°C—548.5μS/cm
Specifications
Packaging Polyethylene
CAS 7447-40-7
Density 1.00g/mL
Conductivity 500μS/cm
Molecular Formula ClK
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
Quantity 500 mL
ChEBI CHEBI:32588
Formula Weight 74.56g/mol
Chemical Name or Material Potassium Chloride