Learn More
Conductivity Standard, 1413μS/cm, NIST Certified, Chem-Lab
Product Code. 11920339
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1 L
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11920339 500 mL Each
11953083 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11920339 Supplier Chem-Lab Supplier No. CL03.1101.0500
  • ONLINE EXCLUSIVE PRICE
    296.00 NOK
    231.19 NOK / Pack of 1
    Save 64.81 NOK
    21% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Calibrates electrodes and meters for accurate electrical and thermal conductivity measurements. Conductivity Standard, 1413μS/cm, NIST Certified, Chem-Lab is a 0.01M potassium chloride solution for laboratory use and LF measurements. 1278μS at 20°C, 1413μS at 25°C (±1%).
5°C—896μS/cm
10°C—1020μS/cm
15°C—1147μS/cm
20°C—1278μS/cm
25°C—1413μS/cm
30°C—1552μS/cm
35°C—1694μS/cm
40°C—1839μS/cm
Chemical Identifiers
CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]
Specifications
Packaging Polyethylene
Density 1.01g/mL
Conductivity 1413μS/cm
Concentration 0.01 M
IUPAC Name potassium;chloride
Quantity 500 mL
Formula Weight 74.56g/mol
Chemical Name or Material Potassium Chloride