Learn More
Conductivity Standard, 100μS/cm, NIST Certified, Chem-Lab
Product Code. 11983083
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11983083 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11983083 Supplier Chem-Lab Supplier No. CL03.1119.0500
  • ONLINE EXCLUSIVE PRICE
    466.00 NOK
    364.48 NOK / 500mL
    Save 101.52 NOK
    21% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Calibrates electrodes and meters for accurate electrical and thermal conductivity measurements. Conductivity Standard, 100μS/cm, NIST Certified, Chem-Lab is a potassium chloride solution for laboratory use and LF measurements. 100μS (±1%) at 25°C.
15°C—80.56μS/cm
16°C—82.50μS/cm
17°C—84.45μS/cm
18°C—86.39μS/cm
19°C—88.33μS/cm
20°C—90.28μS/cm
21°C—92.22μS/cm
22°C—94.17μS/cm
23°C—96.11μS/cm
24°C—98.06μS/cm
25°C—100.00μS/cm
30°C—109.72μS/cm
Specifications
Packaging Polyethylene
CAS 7447-40-7
Density 1.00g/mL
Conductivity 100μS/cm
Molecular Formula ClK
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium;chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
Quantity 500 mL
ChEBI CHEBI:32588
Formula Weight 74.56g/mol
Chemical Name or Material Potassium Chloride