Learn More
Conductivity Standard Solution, 7000μS/cm, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Traceability to NIST | Yes |
| Conductivity | 7000μS/cm |
| Physical Form | Liquid |
| Quantity | 500 mL |