Learn More
Conductivity Standard at 25°C, NIST Traceable, 500,000 μS/cm, Reagecon™
Product Code. 16532350 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16532350 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16532350 Supplier Reagecon Supplier No. CSKC500M10L
 Restricted Item
Estimated Shipment: 19-10-2026
Tested and certified by an ISO17025 Accredited Test Method (A2LA Ref: 6739.03) and NIST traceable.
  • Reagecon offers high quality Conductivity Standards that support accurate conductivity measurements and can be used for sensor calibration, quality control, method validation, and instrument qualification.
  • The standards are aqueous-based, minimising errors caused by matrix mismatch.
  • Directly traceable to NIST reference materials, meeting the specification requirements of all major Pharmacopoeias.
  • Provided with a comprehensive Certificate of Analysis.
  • A temperature conductivity table can be accessed via the product label and clearly displayed lot numbers and expiry dates for ease of use.
  • A low temperature coefficient reduces temperature related errors, allowing reliable measurements without strict temperature control and supporting field use.
  • High accuracy allows the standards function effectively as both calibrators and controls for demanding industrial applications.
  • Manufactured to stringent specifications, ensuring extended shelf life and stability.
Specifications
Chemical Name or Material Conductivity Standard
Packaging HDPE Container
Conductivity 500,000 μS/cm ±1.0%
Type Conductivity Standard
Molecular Weight (g/mol) Unknown
Quantity 10 L
Grade Certified
compliance-icons compliance-icons
Product Identifier
  • CONDUCTIVITY STANDARD 500,000µS/cm @ 25°C Certified Traceable to N.I.S.T.
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid