Learn More
Basic Line Conductivity Standard, 5μS/cm, Hamilton™
Product Code. 10747413
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10747413 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10747413 Supplier Hamilton Supplier No. 238926
  • ONLINE EXCLUSIVE PRICE
    2010.87 NOK
    1883.01 NOK / Pack of 1
    Save 127.86 NOK
    6% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Provides unprecedented stability. Hamilton™ Basic Line Conductivity Standard, 5μS/cm comes with a certificate with calibration document form DFM.
  • Remains stable for up to 3 years
  • Expiration date is shown on every bottle
  • Bottles are permitted to stay open for a total of 60 minutes
Specifications
Packaging Glass bottle
Quantity 250 mL