Description
- Get accurate, reliable measurements with these replacement probes and adapters.
- Features a conductivity range of 1.0 to 200,000 microsiemens
- Suitable for use with 19601-06 meter
Specifications
Specifications
| Type | Conductivity Probe |
| Certifications/Compliance | None |
| For Use With (Equipment) | Traceable High Accuracy, Benchtop Conductivity Meter and Probe with Calibration (19601-06) |