Learn More
WTW™ Conductivity kit
Specifications
Specifications
| Accuracy (TDS) | ±2% |
| Description | Conductivity kit |
| Includes | 6R/SET/Lab1, LR 325/01 ultrapure water cond cell, D 01/T glass flow-through vessel |
| Resolution (Conductivity) | 5μS/cm |
| For Use With (Equipment) | Conductivity meter series |