Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with Olives
Product Code. 10171901
Change view
Click to view available options
Length (Metric):
400 mm
500 mm
750 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10171901 750 mm Each
10326171 400 mm Each
10428792 500 mm Each
3 options
Product Code. 10171901
Brand: Lenz Laborglasintrumente 05210704
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Liebig condenser acc. to DIN 12576 and in addition to DIN
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Length (Metric) 750 mm
Standard Taper Joint No NS 29/32
Material Borosilicate Glass (3.3)