Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with GL Threads
Product Code. 10673755
Click to view available options
Length (Metric) Jacket:
400 mm
500 mm
750 mm
Unit Size:
Each
Product Code. 10673755
Brand: Lenz Laborglasintrumente 05210413
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Liebig condensers acc. to DIN 12576 and in addition to DIN
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Includes GL 14 threads with two threaded plastic olives
Length (Metric) Jacket 400 mm
Length (Metric) 400 mm
Quantity 1 Ea.
Standard Taper Joint No NS 24/29