Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with GL Threads
Product Code. 10067989
Change view
Click to view available options
Length (Metric) Jacket:
400 mm
500 mm
750 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Jacket unitSize
10067989 750 mm Each
10882811 500 mm Each
10673755 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10067989 Supplier Lenz Laborglasintrumente Supplier No. 05210714
  • ONLINE EXCLUSIVE PRICE
    1450.00 NOK
    1203.42 NOK / Pack of 1
    Save 246.58 NOK
    17% Off
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Liebig condensers acc. to DIN 12576 and in addition to DIN
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Includes GL 14 threads with two threaded plastic olives
Length (Metric) Jacket 750 mm
Length (Metric) 750 mm
Quantity 1 Ea.
Standard Taper Joint No NS 29/32