Learn More
Lenz Laborglasintrumente Condensers Acc. to Davies, with GL Thread
Product Code. 11566794
Change view
Click to view available options
Standard Taper Joint No:
NS 24/29
NS 29/32
Length (Metric):
250 mm
400 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Unit Size
11566794 NS 29/32 400 mm Each
11546794 NS 29/32 250 mm Each
11556794 NS 24/29 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11566794 Supplier Lenz Laborglasintrumente Supplier No. 05215414
  • ONLINE EXCLUSIVE PRICE
    2125.00 NOK
    1616.92 NOK / Pack of 1
    Save 508.08 NOK
    23% Off
Estimated Shipment: 19-01-2027
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Davies condensers for condensing vapours
  • With NS socket and NS cone
  • With double sized cooling area
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Includes Two threaded plastic olives
Length (Metric) 400 mm
Standard Taper Joint No NS 29/32
Material Duran Glass