Learn More
Lenz Laborglasintrumente Condensers Acc. to Davies
Product Code. 10254101
Click to view available options
Standard Taper Joint No:
NS 24/29
NS 29/32
Length (Metric):
250 mm
400 mm
Unit Size:
Each
Product Code. 10254101
Brand: Lenz Laborglasintrumente 05215403
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Davies condensers for condensing vapours
  • With NS socket and NS cone
  • With double sized cooling area
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Includes Glass olives
Length (Metric) 400 mm
Standard Taper Joint No NS 24/29
Material Borosilicate Glass (3.3)