Learn More
Lenz Laborglasintrumente Condensers acc. to Allihn
Product Code. 10612995
Change view
Click to view available options
Standard Taper Joint No:
NS 14/23
NS 19/26
NS 24/29
Length (Metric):
250 mm
400 mm
Unit Size:
Each
Product Code. Standard Taper Joint No Length (Metric) unitSize
10612995 NS 24/29 400 mm Each
10401834 NS 24/29 250 mm Each
10090654 NS 14/23 250 mm Each
11973421 NS 19/26 250 mm Each
4 options
Product Code. 10612995
Brand: Lenz Laborglasintrumente 05223413
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Allihn condensers acc. to DIN 12581
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Includes Two threaded plastic olives, GL threads 14
Length (Metric) 400 mm
Standard Taper Joint No NS 24/29