Learn More
Lenz Laborglasintrumente Condensers Acc. to Allihn
Product Code. 10293951
Change view
Click to view available options
Standard Taper Joint No:
NS 19/26
NS 24/29
NS 29/32
No. of Bulbs:
5
6
8
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No No. of Bulbs unitSize
10293951 NS 24/29 5 Each
11856204 NS 29/32 6 Each
10162091 NS 24/29 6 Each
10203911 NS 19/26 6 Each
10254051 NS 24/29 8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10293951 Supplier Lenz Laborglasintrumente Supplier No. 05223303
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Allihn condensers acc. to DIN 12581
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Includes Glass olives
Length (Metric) 250 mm
No. of Bulbs 5
Standard Taper Joint No NS 24/29
Material Borosilicate Glass (3.3)