Learn More
Lenz Laborglasintrumente Condensers Acc. to Allihn
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05223202
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10203911
  • 708.08 NOK / Each
Estimated Shipment: 23-07-2024
Description
  • Allihn condensers acc. to DIN 12581
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Specifications
Glass olives
6
160 mm
NS 19/26
Borosilicate Glass (3.3)