Learn More
Concentrate, To Make 1 ppm Arsenic Solution, According to Chinese Pharmacopoeia (ChP), Reagecon
Shop All Reagecon Products
Specifications
Specifications
| Packaging | HDPE bottle |
| Physical Form | Liquid |
| Concentration | 1 ppm |
| Quantity | 100 mL |