missing translation for 'onlineSavingsMsg'
Learn More

Concanavalin A from Canavalia Ensiformis Seeds (Jack Beans), MP Biomedicals

Product Code. 11482082 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11482082

Brand: MP Biomedicals 02150710.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  • Lectin are used in a wide variety of applications in vitro, including: blood grouping and erythrocyte polyagglutination studies, Mitogenic stimulation of lymphocytes, Lymphocyte subpopulation studies, Fractionation of cells and other particles, Histochemical studies of normal and pathological conditions
  • Con-A has been used to elucidate structural changes in the membrane surface of transformed cells
  • Con-A conjugates of antitumor drugs have been used in drug delivery systems in cultured cells

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100mg
Product Type CONCANAVALIN A from Canavalia ensiformis seeds (jack beans)
Content And Storage -20°C
Form Powder
Color White
Solubility Soluble in water (10 mg/ mL-slightly hazy, colorless solution).
compliance-icons compliance-icons
Product Identifier
  • Concanavalin A
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Concanavalin A
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.