Learn More
Cole-Parmer™ Compression to Male Threaded Adapter
Click to view available options
Port Size:
1/2 in. NPT(M)
3/4 in. NPT(M)
3/8 in. NPT(M)
Unit Size:
Each
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- PVDF Composition
Specifications
Specifications
| Description | Compression x NPT(M) Connection |
| Autoclavable | Autoclavable |
| Material | PVDF |
| Overall Length (English) | 1 15/16 in. |
| Packaging Quantity | 10 Fittings |
| Tubing Size | 3/4 in. |
| Outlet Port | 3/4 in. |
| Port Size | 3/4 in. NPT(M) |
| Sterilization Method | Autoclave or Ethylene Oxide |
| Width (Metric) | 33 mm |
| Show More |