Learn More
Compound E (gamma-Secretase Inhibitor XXI), TRC
Specifications
Specifications
| Chemical Name or Material | Compound E (gamma-Secretase Inhibitor XXI) |
| CAS | 209986-17-4 |
| Molecular Formula | C27H24F2N4O3 |
| Quantity | 0.5 mg |
| Synonym | N-[(1S)-2-[[(3S)-2,3-dihydro-1-methyl-2-oxo-5-phenyl-1H-1,4-benzodiazepin-3-yl]amino]-1-methyl-2-oxoethyl]-3,5-difluorobenzeneacetamide |
| InChI Formula | InChI=1S/C27H24F2N4O3/c1-16(30-23(34)14-17-12-19(28)15-20(29)13-17)26(35)32-25-27(36)33(2)22-11-7-6-10-21(22)24(31-25)18-8-4-3-5-9-18/h3-13,15-16,25H,14H2,1-2H3,(H,30,34)(H,32,35)/t16-,25+/m0/s1 |
| SMILES | C[C@H](NC(=O)Cc1cc(F)cc(F)c1)C(=O)N[C@H]2N=C(c3ccccc3)c4ccccc4N(C)C2=O |
| IUPAC Name | (2S)-2-[[2-(3,5-difluorophenyl)acetyl]amino]-N-[(3S)-1-methyl-2-oxo-5-phenyl-3H-1,4-benzodiazepin-3-yl]propanamide |
| Molecular Weight (g/mol) | 490.5 |
| Formula Weight | 490.1816 |