Learn More
Epredia™ Complete Manual Staining Assemblies 117 and 130
Accommodate up to 60 slides with Epredia™ Complete Manual Staining Assemblies 117 and 130, available with a non-autoclavable glass or stainless-steel dish.
Brand:  Epredia™ 132
Additional Details : Weight : 0.27900kg
 View more versions of this product
Product Code. 12647716
  • 473.00 NOK / Each
Estimated Shipment: 21-06-2024
Description
Complete Manual Staining Assembly 117:
  • Staining Dish 119: Stainless steel with inside dimensions of 7.95 x 3.35 x 2.95″ (202 x 85 x 75mm); holds up to 750mL of solution with fully loaded staining rack.
  • Staining Rack 120: Stainless steel; holds up to 60 slides; maximum slide size is 3 x 2″ (75 x 50mm).
  • Staining Dish Cover 118: Stainless steel
Complete Manual Staining Assembly 130:
  • Staining Dish 131: Nonautoclavable glass with inside dimensions of 7.95 x 4.4 x 2.8″ (202 x 112 x 72mm); holds up to 750mL of solution with fully loaded staining rack
  • Staining Rack 120: Stainless steel; holds up to 60 slides; maximum slide size is 3 x 2″ (75 x 50mm)
  • Staining Dish Cover 132: Stainless steel
Specifications
Specifications
Staining Dish Cover
Staining Dish Cover Only (Assembly 121)