Learn More
SI Analytics™ Complete Dosing Unit
Product Code. 10267871 Shop All SI Analytics Products
Change view
Click to view available options
Volume (Metric):
20 mL
50 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10267871 20 mL Each
10499821 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10267871 Supplier SI Analytics™ Supplier No. 285223718
  • ONLINE EXCLUSIVE PRICE
    3860.00 NOK
    3621.18 NOK / Pack of 1
    Save 238.82 NOK
    6% Off
 Restricted Item
In Stock
SI Analytics™ Complete Dosing Unit comes with piston and piston rod, as well as a UV protection coat for use with the TITRONIC™ Series.
  • Dosing unit
  • Includes piston, piston rod, and UV protection coat
  • For use wtih TITRONIC™ Universal and TITRONIC™ 300
TRUSTED_SUSTAINABILITY
Specifications
Type Complete Dosing Unit
Includes Piston, Piston Rod and UV Protection Coat
Volume (Metric) 20 mL