Learn More
Nabertherm™ Compact Tube Furnace with C550 Controller
Product Code. p-7236256 Shop All Nabertherm Products
Change view
Click to view available options
Diameter (Metric) Process Tube:
50 mm
120 mm
170 mm
Heated Zone:
250 mm
500 mm
750 mm
1000 mm
Temperature (Metric):
1200°C
1300°C
Unit Size:
Each
Product Code. Diameter (Metric) Process Tube Heated Zone Temperature (Metric) unitSize
17338079 50 mm 250 mm 1200°C Each
17348079 50 mm 250 mm 1300°C Each
17378079 50 mm 500 mm 1200°C Each
17388079 50 mm 500 mm 1300°C Each
17338089 120 mm 500 mm 1200°C Each
17348089 120 mm 500 mm 1300°C Each
17398089 170 mm 750 mm 1200°C Each
17308099 170 mm 750 mm 1300°C Each
17358099 170 mm 1000 mm 1200°C Each
17368099 170 mm 1000 mm 1300°C Each
10 options
Product Code. 17338079
Brand: Nabertherm™ R152O2AN
Only null left
Add to basket
Add to basket
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
  • Excellent price - performance ratio
  • Can also be equipped with gas supply system 1 (no vacuum operation) and gas supply system 2 (for vacuum operation)
  • Temperature, max. 1200°C or 1300°C
  • Five programs and four freely programmable ramps, controller C 550 with 10 freely adjustable programs and 20 segments
  • Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
  • Dual shell housing made of sheets of textured stainless steel
  • Insulation made of non-classified fibre material
Specifications
Controller Type C550
Heated Zone 250 mm
Temperature (Metric) 1200°C
Type Compact Tube Furnace
Dimensions (L x W x H) Exterior 434 x 340 x 508 mm
Length (Metric) 450 mm
Diameter (Metric) Process Tube 50 mm