Learn More
Hanna Instruments™ Compact Thermohygrometer
Specifications
Specifications
| Type | Compact Thermohygrometer |
| Resolution | 0.1°C |
| Temperature Range (Metric) | 0°C to 60°C |
| Temperature Accuracy | ±3°C |
| Temperature Resolution | 0.1°C |
| Weight (Metric) | 340 g |
| Electrical Requirements | 9V |
| Voltage | 9 V |
| Includes | Dew Point, Calib. Data-Log Probe |