Learn More
Kern Sohn Compact Laboratory Balance PCB
Click to view available options
Capacity:
100 g
350 g
1000 g
3500 g
Unit Size:
Each
Description
- PRE-TARE function for manual subtraction of a known container weight, useful for checking fill-levels
- Freely programmable weighing unit, e.g. display direct in special units such as length of thread g/m, paper weight g/m2, or similar
- With the recipe function you can weigh the different ingredients of a mixture. As a check, you can also call up the total weight of all the ingredients
- Plus/minus weighing procedures
- Protective working cover included with delivery
Specifications
Specifications
| Capacity | 100 g |
| Readability | 0.001 g |
| Linearity | 0.003 g |
| Repeatability | 0.001 g |
| Type | Compact Precision Blanace |
| Length (Metric) Pan | 81 mm |
| Width (Metric) Pan | 81 mm |
| Electrical Requirements | Universal Mains Adapter |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.