Learn More
Thermo Scientific™ Lid
Material: Plastic; For Use With: Thermo Scientific™ Compact D Model Dry Bath/Block Heater
Brand: Thermo Scientific™ 88871111
Additional Details : Weight : 0.50000kg
Description
Increase your application capabilities with a variety of accessories for the Thermo Scientific™ Compact Dry Baths/Block Heaters.
Warranty: 2 yearsSpecifications
Plastic Lid for Compact D models | |
LED | |
Lid |
Thermo Scientific™ Compact Dry Bath/Block Heater | |
Plastic | |
122 x 158 x 30 mm (4.8 x 6.2 x 1.2 in.) |