Learn More
Thermo Scientific™ Block, 3 x 50 mL
Product Code. 15302235 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Places:
3
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Places Unit Size
15302235 3 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15302235 Supplier Thermo Scientific™ Supplier No. 88871106
  • ONLINE EXCLUSIVE PRICE
    1060.00 NOK
    974.51 NOK / Pack of 1
    Save 85.49 NOK
    8% Off
 Restricted Item
In Stock
For Use With: Thermo Scientific™ Compact Dry Bath/Block Heater
Increase your application capabilities with a variety of accessories for the Thermo Scientific™ Compact Dry Baths/Block Heaters.
Warranty: 2 years
TRUSTED_SUSTAINABILITY
Specifications
Description 3 x 50mL block
Capacity (Metric) 50 mL
For Use With (Equipment) Thermo Scientific™ Compact Dry Bath/Block Heater
Lighting LED
No. of Places 3
Type Dry Block
Dimensions (L x W x H) 36 x 108 x 72 mm (1.4 x 4.3 x 2.8 in.)