Learn More
Tenak™ Comfort Rack for Microtiter Plates for Upright Freezers, 65mm Shelf Height
Product Code. p-8543044 Shop All TENAK Products
Click to view available options
Holds:
Plates (10)
Plates (12)
Plates (14)
Plates (16)
Plates (21)
Plates (24)
Plates (28)
Plates (30)
Plates (35)
Plates (36)
Plates (8)
Dimensions (L x W x H):
360 x 135 x 133 mm
360 x 135 x 200 mm
360 x 135 x 266 mm
450 x 135 x 133 mm
450 x 135 x 299 mm
540 x 135 x 133 mm
540 x 135 x 266 mm
540 x 135 x 332 mm
540 x 135 x 399 mm
630 x 135 x 133 mm
630 x 135 x 200 mm
630 x 135 x 266 mm
630 x 135 x 332 mm
Unit Size:
Each
Product Code. 12407070
Brand: Tenak™ TE35508
Only null left
Add to basket
Add to basket
Providing a complete range of racks for microtiter plates in chest freezers. Tenak™ Comfort Racks for Microtiter Plates in Upright Freezers are stackable.
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • With an alert preventing the shelves from sliding out
  • External footprint is 135 x 92mm
Specifications
Product Type Microtiter Plate Rack
Array 6 x 5
Holds Plates (30)
Shelves 30
Stackable Yes
For Use With (Equipment) Microtiter Plates, Upright Freezers
No. per Pack 1 Pack
Dimensions (L x W x H) 450 x 135 x 299 mm
Width (Metric) 135 mm
Height (Metric) 299 mm
Height (Metric) Shelf 65 mm
Depth (Metric) 450 mm