Learn More
Tenak™ Comfort Rack for Microtiter Plates for Upright Freezers, 25mm Shelf Height
Product Code. 12497050 Shop All TENAK Products
Change view
Click to view available options
Holds:
Plates (24)
Plates (30)
Plates (32)
Plates (36)
Plates (40)
Plates (42)
Plates (48)
Plates (50)
Plates (60)
Plates (70)
Plates (72)
Plates (75)
Plates (84)
Dimensions (L x W x H):
360 x 135 x 159 mm
360 x 135 x 212 mm
360 x 135 x 265 mm
360 x 135 x 318 mm
450 x 135 x 159 mm
450 x 135 x 212 mm
450 x 135 x 265 mm
450 x 135 x 318 mm
450 x 135 x 371 mm
450 x 135 x 398 mm
540 x 135 x 159 mm
540 x 135 x 212 mm
540 x 135 x 265 mm
540 x 135 x 318 mm
630 x 135 x 159 mm
630 x 135 x 318 mm
Unit Size:
Each
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Dimensions (L x W x H) unitSize
12497050 Plates (75) 450 x 135 x 398 mm Each
12407080 Plates (60) 540 x 135 x 265 mm Each
12417080 Plates (72) 540 x 135 x 318 mm Each
12427100 Plates (84) 630 x 135 x 318 mm Each
12447050 Plates (30) 450 x 135 x 159 mm Each
12457050 Plates (40) 450 x 135 x 212 mm Each
12467030 Plates (24) 360 x 135 x 159 mm Each
12467050 Plates (50) 450 x 135 x 265 mm Each
12477030 Plates (32) 360 x 135 x 212 mm Each
12477050 Plates (60) 450 x 135 x 318 mm Each
12487030 Plates (40) 360 x 135 x 265 mm Each
12487050 Plates (70) 450 x 135 x 371 mm Each
12487070 Plates (36) 540 x 135 x 159 mm Each
12497030 Plates (48) 360 x 135 x 318 mm Each
12497070 Plates (48) 540 x 135 x 212 mm Each
12497090 Plates (42) 630 x 135 x 159 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 12497050 Supplier Tenak™ Supplier No. TE35470
Providing a complete range of racks for microtiter plates in chest freezers. Tenak™ Comfort Racks for Microtiter Plates in Upright Freezers are stackable.
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • With an alert preventing the shelves from sliding out
  • External footprint is 135 x 92mm
Specifications
Holds Plates (75)
Stackable Yes
For Use With (Equipment) Microtiter Plates, Upright Freezers
No. per Pack 1 Pack
Dimensions (L x W x H) 450 x 135 x 398 mm
Height (Metric) Shelf 25 mm
Depth (Metric) 450 mm
Product Type Microtiter Plate Rack
Shelves 75
Height (Metric) 398 mm
Array 15 x 5
Width (Metric) 135 mm