Learn More
R&D Systems™ CometAssay Electrophoresis System
Product Code. 30001322
Change view
Click to view available options
Content And Storage:
The product requires storage at both 2 - 8°C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
30001322 The product requires storage at both 2 - 8°C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Caution! Electrical Hazard! This equipment is designed for use with a DC power supply providing up to 250 VDC. Careless handling can result in electrical shock. To avoid any risk of injury, the instrument should be operated in accordance with the instruction provided.
Product Code. 30001322 Supplier R&D Systems™ Supplier No. 4250050ES
  • ONLINE EXCLUSIVE PRICE
    36796.26 NOK
    32560.50 NOK / 1 sets
    Save 4235.76 NOK
    11% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Caution! Electrical Hazard! This equipment is designed for use with a DC power supply providing up to 250 VDC. Careless handling can result in electrical shock. To avoid any risk of injury, the instrument should be operated in accordance with the instruction provided.
Electrophoresis System for the CometAssay™
  • Overcomes assay variability first by placing an acrylic overlay on top of an elevated slide tray to maintain optimal buffer height for DNA migration
  • Secondly, a constant buffer temperature is maintained using an external chamber under the tank to cool the glass slide platform and buffer chamber
  • Electrophoresis time for optimal DNA migration is achieved in 30 minutes with Alkaline (pH>13) Electrophoresis Solution, or 45 minutes with Neutral Electrophoresis Buffer at 1V/cm
  • The smoke grey colored unit is designed to minimize exposure to UV light but still allow visual inspection
  • Third, specially designed slide trays are also provided to accommodate 2, 20 and 96 well slides and maintain proper slide orientation during electrophoresis
  • CometAssay™ Electrophoresis System II, CometAssay™ Alkaline Control Cells, and CometAssay™ Neutral Control Cells enable investigators to consistently optimize their comet assays for highly reproducible results, and to standardize electrophoresis methods between individual users and laboratories
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage The product requires storage at both 2 - 8°C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
For Use With (Application) A simple and effective method for evaluating DNA damage in cells
Includes Electrophoresis Tank with Leveling Bubble, Safety Lid & Cables, 2/20 Well Slide Trays, and Slide Tray Overlay
Product Line R&D Systems