Learn More
Fisherbrand™ Combs for TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis Unit
Click to view available options
No. of Wells:
8
12
15
20
Volume (Metric) Sample:
10 μL
15 μL
20 μL
25 μL
35 μL
45 μL
50 μL
65 μL
90 μL
Width (Metric) Teeth:
2.5 mm
4 mm
5.5 mm
9.25 mm
Unit Size:
Each
Description
- Available in 1, 1.5 and 2mm thickness
Specifications
Specifications
| For Use With (Equipment) | TVG-SYS Vari-Gel MIDI Horizontal Gel Electrophoresis Unit |
| Type | Comb |
| No. of Wells | 20 |
| Thickness (Metric) | 2 mm |
| Volume (Metric) Sample | 20 μL |
| Width (Metric) Teeth | 2.5 mm |