Learn More
Fisherbrand™ Combs for TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis Unit
Product Code. 11726185
Change view
Click to view available options
No. of Wells:
8
12
15
20
Volume (Metric) Sample:
10 μL
15 μL
20 μL
25 μL
35 μL
45 μL
50 μL
65 μL
90 μL
Width (Metric) Teeth:
2.5 mm
4 mm
5.5 mm
9.25 mm
Unit Size:
Each
Product Code. No. of Wells Volume (Metric) Sample Width (Metric) Teeth unitSize
11726185 8 65 μL 9.25 mm Each
11736185 12 35 μL 5.5 mm Each
11796185 20 15 μL 2.5 mm Each
11716175 8 45 μL 9.25 mm Each
11706195 8 90 μL 9.25 mm Each
11706185 12 25 μL 5.5 mm Each
11724925 15 20 μL 4 mm Each
11716185 20 10 μL 2.5 mm Each
11716195 12 50 μL 5.5 mm Each
11726195 20 20 μL 2.5 mm Each
10 options
Product Code. 11726185
Brand: Fisherbrand™ TVGC158
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis Unit
  • Available in 1, 1.5 and 2mm thickness
Specifications
For Use With (Equipment) TVG-SYS Vari-Gel MIDI Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 8
Thickness (Metric) 1.5 mm
Volume (Metric) Sample 65 μL
Width (Metric) Teeth 9.25 mm