Learn More
Fisherbrand™ Combs for HU13 Midi Horizontal Gel Electrophoresis Unit, 2mm thick
Product Code. 12358426
Click to view available options
No. of Wells:
10
12
16
24
28
Width (Metric) Teeth:
3 mm
5.5 mm
8 mm
9.5 mm
Volume (Metric) Sample:
25 μL
50 μL
70 μL
85 μL
Unit Size:
Each
Product Code. 12358426
Brand: Fisherbrand™ HU13C212MC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with HU13 Midi Horizontal Gel Electrophoresis Unit
  • Color coded
  • Height adjustable
  • Offer complete control over loading volume
  • Well depth to maximum 112-sample throughput
Specifications
For Use With (Equipment) HU13 Midi Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 12
Thickness (Metric) 2 mm
Volume (Metric) Sample 70 μL
Width (Metric) Teeth 8 mm