Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 0.75mm Thick
For use in preparatory electrophoresis and high-throughput screening.
227.00 NOK - 252.00 NOK
Specifications
Type Comb
Quantity 1 Each
Thickness (Metric) 0.75 mm
View More Specs
Products 9
Product Code Brand No. of Wells Volume (Metric) Sample Price Quantity & Availability  
Product Code Brand No. of Wells Volume (Metric) Sample Price Quantity & Availability  
11883303
View Documents Promotion Details
Fisherbrand™
1 Prep, 1 Marker 270 μL
227.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
11893303
View Documents Promotion Details
Fisherbrand™
10 20 μL
227.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
11863313
View Documents Promotion Details
Fisherbrand™
12 17 μL
227.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
11803323
View Documents Promotion Details
Fisherbrand™
16 12 μL
227.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
11843323
View Documents Promotion Details
Fisherbrand™
2 Prep, 2 Marker 57 μL
227.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
11853323
View Documents Promotion Details
Fisherbrand™
20 10 μL
227.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
11823333
View Documents Promotion Details
Fisherbrand™
25 7 μL
252.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
11863333
View Documents Promotion Details
Fisherbrand™
4 Prep, 2 Marker 118 μL
227.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
11803343
View Documents Promotion Details
Fisherbrand™
8 30 μL
227.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
Description
  • For tightly resolved bands
  • Recommended for high resolution gels and publication quality data
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Comb
1 Each
0.75 mm