Learn More
Fisherbrand™ Combs for CHU25 Maxi-Plus Cooled Horizontal Gel Electrophoresis Unit
For use with CHU25 Maxi-Plus Cooled Horizontal Gel Electrophoresis Unit
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Color coded
- Height adjustable
- Offer complete control over loading volume and well depth
Specifications
CHU25 Maxi-Plus Cooled Horizontal Gel Electrophoresis Unit | |
26 | |
30 μL |
Comb | |
1 mm | |
7 mm |