Learn More
Lovibond™ Combi pack CHLORINE HR / ACIDIFYING GP
Tablet reagent test set
Brand:  Lovibond™ 517721BT
Additional Details : Weight : 0.07020kg
Product Code. 15351925
  • 840.00 NOK / Pack of 100
Estimated Shipment: 14-06-2024
Description
Tablet reagent test set
Combi pack CHLORINE HR / ACIDIFYING GP 100 tests measuring range 0.2 - 16 % NaOCl Potassium iodide method for comparators and photometers.
Specifications
Specifications
100 Tests
CHLORINE 0.2 to 16 %
Combi Pack Chlorine HR/Acidifying GP
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • combi-pack chlorine HR + acidifying GP
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : citric acid, adipicacid