Learn More
Macherey-Nagel™ NucleoSpin™ RNA Plus, Columns for RNA purification with DNA removal column
Product Code. 15820978
Change view
Click to view available options
Quantity:
50
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15820978 50 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15820978 Supplier Macherey-Nagel™ Supplier No. 740984.50S
  • ONLINE EXCLUSIVE PRICE
    2525.00 NOK
    2290.91 NOK / Pack of 50
    Save 234.09 NOK
    9% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Mini spin kit for ultrafast and convenient RNA isolation without rDNase digestion
NucleoSpin™ RNA Plus
  • Efficient and quick RNA isolation from cells and tissues
  • Two-column system includes gDNA removal columns and RNA binding columns
  • NucleoSpin™ gDNA Removal Columns – no time consuming rDNase digestion
  • Convenient handling – lysate clearing and gDNA removal on one column in one step
  • New efficient lysis buffer – no b-mercaptoethanol or TCEP necessary
  • Variety of sample materials – cultured cells, animal tissue, plant tissue, microorganisms
  • Downstream applications include real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays
Specifications
Color Blue ring
Content And Storage Room temperature (18-25 °C)
Format Mini Spin Column
Isolation Technology Silica Membrane
Elution Volume 30 - 120 μL
Sample Type Cultured Bacterial or Yeast Cells, Human or Animal Tissue
Efficiency Binding capacity: 200 μg
Final Product Type RNA (with typical RIN > 9)
For Use With (Equipment) NucleoSpin™ RNA Plus Kits
For Use With (Application) Isolation of RNA
Product Type RNA Plus Binding Column
Quantity 50
Sample Size <1 x 107 cultured cells, <30 mg human/animal tissue, <109 bacterial cells, <108 yeast cells,<1 x 107 cultured cells, <30 mg human/animal tissue, <109 bacterial cells, <108 yeast cells,<1 x 107 cultured cells, <30 mg human/animal tissue, <109 bacterial cells, <108 yeast cells
Test Time 20 min/6 preps
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • NucleoSpin RNA Plus
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: ethyleneglycol diacetate / guanidinium thiocyanate /guanidine hydrochloride, ethanol