Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel with Removable Glass Jacket
Product Code. 10719411
Change view
Click to view available options
Length (Metric):
200 mm
300 mm
500 mm
800 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10719411 200 mm Each
10000551 500 mm Each
10010551 800 mm Each
10498402 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10719411 Supplier Lenz Laborglasintrumente Supplier No. 05117201
  • ONLINE EXCLUSIVE PRICE
    1140.00 NOK
    942.79 NOK / Pack of 1
    Save 197.21 NOK
    17% Off
Estimated Shipment: 28-12-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Columns acc. to Hempel
  • With removable glass jacket
  • For packings like Raschig rings or glass beads
  • With NS socket and NS cone
  • With removable glass jacket
  • Made of DURAN™ tubing
Specifications
Includes Removable glass jacket
Length (Metric) 200 mm
Standard Taper Joint NS 14/23