Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel with Removable Glass Jacket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05117301
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10498402
  • 1065.30 NOK / Each
Estimated Shipment: 08-07-2024
Description
  • Columns acc. to Hempel
  • With removable glass jacket
  • For packings like Raschig rings or glass beads
  • With NS socket and NS cone
  • With removable glass jacket
  • Made of DURAN™ tubing
Specifications
Specifications
Removable glass jacket
300 mm
NS 14/23