Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel
Product Code. p-7222631
Change view
Click to view available options
Standard Taper Joint:
Cone and Socket NS 14/23
Cone and Socket NS 24/29
Cone and Socket NS 29/32
Length (Metric):
300 mm
500 mm
800 mm
Unit Size:
Each
Product Code. Standard Taper Joint Length (Metric) unitSize
10141761 Cone and Socket NS 29/32 300 mm Each
10020501 Cone and Socket NS 24/29 800 mm Each
10129310 Cone and Socket NS 14/23 300 mm Each
10224581 Cone and Socket NS 24/29 300 mm Each
10509581 Cone and Socket NS 24/29 500 mm Each
10749031 Cone and Socket NS 29/32 800 mm Each
6 options
Product Code. 10141761
Brand: Lenz Laborglasintrumente 05115304
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Columns acc. to Hempel
  • With removable glass jacket
  • For packings like Raschig rings or glass beads
  • With NS socket and NS cone
  • With removable glass jacket
  • Made of DURAN™ tubing
Specifications
Length (Metric) 300 mm
Standard Taper Joint Cone and Socket NS 29/32