Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel
Product Code. 10020501
Change view
Click to view available options
Standard Taper Joint:
Cone and Socket NS 14/23
Cone and Socket NS 24/29
Cone and Socket NS 29/32
Length (Metric):
300 mm
500 mm
800 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Length (Metric) Unit Size
10020501 Cone and Socket NS 24/29 800 mm Each
10141761 Cone and Socket NS 29/32 300 mm Each
10129310 Cone and Socket NS 14/23 300 mm Each
10224581 Cone and Socket NS 24/29 300 mm Each
10509581 Cone and Socket NS 24/29 500 mm Each
10749031 Cone and Socket NS 29/32 800 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10020501 Supplier Lenz Laborglasintrumente Supplier No. 05115803
  • ONLINE EXCLUSIVE PRICE
    944.00 NOK
    783.77 NOK / Pack of 1
    Save 160.23 NOK
    16% Off
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Columns acc. to Hempel
  • With removable glass jacket
  • For packings like Raschig rings or glass beads
  • With NS socket and NS cone
  • With removable glass jacket
  • Made of DURAN™ tubing
Specifications
Length (Metric) 800 mm
Standard Taper Joint Cone and Socket NS 24/29