Learn More
OHAUS™ Column Mounts
Product Code. 15938465 Shop All Ohaus Products
Change view
Click to view available options
Type:
CS D52xx
SST D52xx
Weight (Metric):
2.300 kg
4.100 kg
6.200 kg
Unit Size:
1 unit
Product Code. Type Weight (Metric) unitSize
15938465 CS D52xx 2.300 kg 1 unit
15948465 CS D52xx 4.100 kg 1 unit
15958465 CS D52xx 6.200 kg 1 unit
15968465 SST D52xx 2.300 kg 1 unit
15978465 SST D52xx 4.100 kg 1 unit
15988465 SST D52xx 6.200 kg 1 unit
6 options
Product Code. 15938465
Brand: OHAUS™ 30424410
OHAUS™ Column Mounts are designed for use with a range of Defender™ 5000 Bases.
  • For use with a base
TRUSTED_SUSTAINABILITY
Specifications
Languages Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX)
Size 35 cm
Weight (Metric) 2.300 kg
Product Line Column Kit
Type CS D52xx