missing translation for 'onlineSavingsMsg'
Learn More

Ansell Colortext Plus F079 Medium Duty Cut-Resistant Knit Gloves

Product Code. 16845380
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves are knitted using a seamless non-stop process with nylon and high performance yarns.

  • Gloves are knitted with a seamless, non-stop process
  • Constructed with nylon and high performances yarns
  • Cotton yarn on the inside of glove provides additional comfort
  • Elastic knitwrist ensures a snug fit
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 407:2004, EN 420:2003 + A1:2009, REACH Compliant
Coating Material Uncoated
Color Navy
Cut Resistance Rating ANSI A2
Material Nylon
Size 8
EN388 Standard 2X4XB
For Use With (Application) Automotive Assembly, Machinery, and Equipment Operation
Glove Cuff Style Knit
Series F079
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'