missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Color-Coded Silicone Vacuum Tubing

Providing tubing solutions for a range of applications, including automotive and industrial uses. Fisherbrand™ Color-Coded Silicone Vacuum Tubing is also useful for low pressure solar installations where tubing is used to circulate water through the solar systems.

Brand:  Fisherbrand™ 1173-3179

 View more versions of this product

Product Code. 11733179

  • 2095.00 NOK / Each
Estimated Shipment: 13-06-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Autoclavable
  • Safe for food use
  • Good for high temperature work
  • Excellent resistance to crushing
  • Flexible, odorless and non-toxic
Specifications

Specifications

FDA, BFR and WRAS approved
9.6 mm
12.8 mm
1.6 mm
-60°C
Flexible
250°C
Autoclavable
Silicone
25 m
Ideal for rapid identification, hot work
Green
1.5 bar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.